L(-)-Xylose 99+%

AC22599-0050 AC22599-0250
200037-740EA 47.7 USD
200037-740 200043-728
L(-)-Xylose 99+%
L(-)-Xylose
Formula: C₅H₁₀O₅
MW: 150.13 g/mol
Melting Pt: 150…152 °C
Storage Temperature: Ambient
MDL Number: MFCD00151096
CAS Number: 609-06-3
EINECS: 210-174-1

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤0.5 %
HPLC ≥99.0 % (ELSD)
Specific optical rotation -18° to -20° (20 °C, 589 nm) (c=10, H2O) (after 24 h)
Loss on ignition ≤0.1 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR