Xylitol 99+%

Synonyms: (2R,3r,4S)-Pentane-1,2,3,4,5-pentaol

AC22598-0025 AC22598-0250 AC22598-1000
103466-772EA 18.2 USD
103466-772 103466-770 200008-066
Xylitol 99+%
Xylitol
Formula: C₅H₁₂O₅
MW: 152.15 g/mol
Boiling Pt: 215…217 °C
Melting Pt: 92…96 °C
Storage Temperature: Ambient
MDL Number: MFCD00064292
CAS Number: 87-99-0
EINECS: 201-788-0
Merck Index: 12,10218

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 92 to 96 °C
Water ≤0.5 %
Sulfated ash ≤0.5 %
GC ≤2.0 % Other polyols
GC ≥99.0 % (on dry substance)
Arsenic (As) ≤3 ppm
Heavy metals (as Pb) ≤10 ppm
Reducing sugars ≤0.2 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR