2-Aminoisobutyric acid 98+%

Synonyms: 2-Amino-2-methylpropanoic acidα-Aminoisobutyric acid2-Amino-2-methylpropionic acid2-Methylalanine2-MetilalaninaH-Aib-OH

AC22576-0250 AC22576-1000
200004-950EA 102 USD
200004-950 200022-440
2-Aminoisobutyric acid 98+%
2-Aminoisobutyric acid
Formula: C₄H₉NO₂
MW: 103.12 g/mol
Boiling Pt: 204 °C (1013 hPa)
Melting Pt: 335 °C
MDL Number: MFCD00008049
CAS Number: 62-57-7
EINECS: 200-544-0
Merck Index: 13,00444

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Titration with HClO4 ≥98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR