(3,4-Dihydroxyphenyl)acetic acid 98%

Warning

Synonyms: DOPAC3,4-Dihydroxyphenylacetic acid

AC22560-0050 AC22560-0250 AC22560-0010
200016-854EA 30.1 USD
200016-854 200016-856 103465-074
(3,4-Dihydroxyphenyl)acetic acid 98%
(3,4-Dihydroxyphenyl)acetic acid
Formula: (C₆H₃(OH)₂)CH₂COOH
MW: 168.15 g/mol
Melting Pt: 126…130 °C
Storage Temperature: Ambient
MDL Number: MFCD00004338
CAS Number: 102-32-9
EINECS: 203-024-1

Order Now

Specification Test Results

Appearance Beige to light brown to purple-grey Powder
Infrared spectrum Conforms
Melting point 127 to 132 °C
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR