Calcium sulfate dihydrate 98+% for analysis

Synonyms: Plaster of Paris

AC22527-0025
103463-496EA 119 USD
103463-496
Calcium sulfate dihydrate 98+% for analysis
Calcium sulphate dihydrate
Formula: CaSO₄·2H₂O
MW: 172.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00149625
CAS Number: 10101-41-4
EINECS: 231-900-3
Merck Index: 13,01711

Order Now

Specification Test Results

Appearance White Powder
Titration Complexometric 98.0 to 102.0 %
Chloride (Cl) ≤50 ppm
Nitrate (NO3) ≤50 ppm
Iron (Fe) ≤20 ppm
Heavy metals (as Pb) ≤20 ppm
Insoluble matter ≤0.05 % (in dilute HCl)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR