4-tert-Butyl-1-bromobenzene 97%

Warning

Synonyms: 1-Bromo-4-(tert-butyl)benzene2-(4-Bromophenyl)-2-methylpropane4-Bromo-tert-butylbenzene1-Bromo-4-tert-butylbenzene

AC22519-1000 AC22519-0050 AC22519-0250
200027-822EA 184 USD
200027-822 200015-722 103511-366
4-tert-Butyl-1-bromobenzene 97%
4-tert-Butyl-1-bromobenzene
Formula: C₁₀H₁₃Br
MW: 213.12 g/mol
Boiling Pt: 80…81 °C (2 mmHg)
Melting Pt: 15…16 °C
Density: 1.22 g/cm³
Flash Pt: 97 °C
Storage Temperature: Ambient
MDL Number: MFCD00000108
CAS Number: 3972-65-4
EINECS: 223-599-2

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5315 to 1.5345 (20 °C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR