2,6-Di-tert-butyl-4-methylpyridine 98%

Warning

Synonyms: 2,6-Di-Tert-butyl-4-methylpyridine

AC22452-0250 AC22452-0050
200046-576EA 145 USD
200046-576 200046-574
2,6-Di-tert-butyl-4-methylpyridine 98%
2,6-Di-tert-butyl-4-methylpyridine
Formula: C₁₄H₂₃N
MW: 205.34 g/mol
Melting Pt: 33…36 °C
Flash Pt: 83 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006305
CAS Number: 38222-83-2
EINECS: 253-834-4
UN: 1325

Order Now

Specification Test Results

Appearance Colorless to yellow to orange to light brown Liquid or low melting solid
Infrared spectrum Conforms
Refractive index 1.4760 to 1.4780 (20 °C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR