(Methyl)triphenylphosphonium iodide 97%

Danger

Synonyms: Methyltriphenylphosphonium iodide

AC22403-0250 AC22403-1000
200019-804EA 61.5 USD
200019-804 200031-976
(Methyl)triphenylphosphonium iodide 97%
(Methyl)triphenylphosphonium iodide
Formula: C₁₉H₁₈IP
MW: 404.23 g/mol
Melting Pt: 185…187 °C
MDL Number: MFCD00066175
CAS Number: 2065-66-9
EINECS: 218-178-5
UN: 3464
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to light yellow Powder
Infrared spectrum Conforms
Melting point 183 to 188 °C
Titration Argentometric ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR