Titanium(IV) ethanolate (33 - 35% TiO₂)

Warning

Synonyms: Titanium (IV) ethoxideTitanium tetraethanolateTetraethyl orthotitanate

AC22376-2500
200058-758EA 153 USD
200058-758
Titanium(IV) ethanolate (33 - 35% TiO₂)
Titanium(IV) ethanolate
Formula: C₈H₂₀O₄Ti
MW: 228.12 g/mol
Boiling Pt: 150…152 °C (10 mmHg)
Melting Pt: –40 °C
Density: 1.09 g/cm³
Flash Pt: 33 °C
Storage Temperature: Ambient
MDL Number: MFCD00009071
CAS Number: 3087-36-3
EINECS: 221-410-8
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5030 to 1.5070 (20 °C, 589 nm)
Titration Complexometric 33 to 35 % (TiO2)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR