2-Hydroxy-p-toluic acid 98%

Warning

Synonyms: 4-Methylsalicylic acidm-Cresotic acidm-Homosalicyclic acid2-Hydroxy-4-methylbenzoic acid

AC22354-1000 AC22354-0250
200029-908EA 50.9 USD
200029-908 200010-570
2-Hydroxy-p-toluic acid 98%
2-Hydroxy-p-toluic acid
Formula: C₈H₈O₃
MW: 152.15 g/mol
Melting Pt: 175…179 °C
MDL Number: MFCD00002454
CAS Number: 50-85-1
EINECS: 200-068-3

Order Now

Specification Test Results

Appearance White to grey-beige Powder
Infrared spectrum Conforms
Melting point 171 to 179 °C
Titration with NaOH ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR