Sodium chlorate 99+%, Extra Pure
|
Danger
|
AC22322-0051
103526-966EA
208
USD
103526-966
Sodium chlorate 99+%, Extra Pure
Sodium chlorate
Formula:
ClNaO₃ MW: 106.44 g/mol Melting Pt: 248…261 °C Density: 2.49 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00003479 CAS Number: 7775-09-9 EINECS: 231-887-4 UN: 1495 ADR: 5.1,II REACH: 01-2119474389-23 Merck Index: 12,08741 |
Specification Test Results
Appearance | White to light yellow Crystalline powder or crystals |
Water | ≤0.05 % |
Titration | ≥99.0 % |
Sodium chloride (NaCl) | ≤1000 ppm |
Insoluble matter | ≤0.005 % (in water) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...