Barium acetate 99+% for analysis

Warning

Synonyms: Barium di(acetate)Acetic acid barium salt

AC22280-0025
103531-912EA 222 USD
103531-912
Barium acetate 99+% for analysis
Barium acetate
Formula: (H₃CCOO)₂Ba
MW: 255.42 g/mol
Melting Pt: ∼450 °C
Density: 2.46 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00012447
CAS Number: 543-80-6
EINECS: 208-849-0
UN: 1564
ADR: 6.1,III
Merck Index: 12,00992

Order Now

Specification Test Results

Appearance White Crystalline powder
Titration Complexometric 99.0 to 102.0 %
pH 6.5 to 8.5 (5 % at 20 °C)
Chloride (Cl) ≤50 ppm
Nitrate (NO3) ≤20 ppm
Calcium (Ca) ≤50 ppm
Iron (Fe) ≤50 ppm
Magnesium (Mg) ≤10 ppm
Potassium (K) ≤30 ppm
Sodium (Na) ≤50 ppm
Strontium (Sr) ≤0.2 %
Heavy metals (as Pb) ≤5 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR