Potassium nitrite 97% for analysis

Danger

AC22270-0050
103526-952EA 461.1 USD
103526-952
Potassium nitrite 97% for analysis
Potassium nitrite
Formula: KNO₂
MW: 85.1 g/mol
Melting Pt: 387 °C
Storage Temperature: Ambient
MDL Number: MFCD00011408
CAS Number: 7758-09-0
EINECS: 231-832-4
UN: 1488
ADR: 5.1,II
Merck Index: 12,07816

Order Now

Specification Test Results

Appearance White to light yellow Crystals
Titration with KMnO4 ≥96.0 %
Chloride (Cl) ≤100 ppm
Sulfate (SO4) ≤200 ppm
Arsenic (As) ≤1 ppm
Iron (Fe) ≤10 ppm
Lead (Pb) ≤10 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR