Lead 99+%, granules

Danger

AC22262-1000 AC22262-5000
200031-698EA 73.34 USD
200031-698 200031-696
Lead 99+%, granules
Lead
Formula: Pb
MW: 207.2 g/mol
Boiling Pt: 1740 °C (1013 hPa)
Melting Pt: 327.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00134050
CAS Number: 7439-92-1
EINECS: 231-100-4
Merck Index: 12,05410

Order Now

Specification Test Results

Appearance Grey Granules
Assay ≥99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR