Dimethyl fumarate 99%

Warning

Synonyms: Fumaric acid dimethyl ester

AC22218-0250 AC22218-5000 AC22218-1000
200012-096EA 27 USD
200012-096 200006-322 103509-744
Dimethyl fumarate 99%
Dimethyl fumarate
Formula: C₆H₈O₄
MW: 144.13 g/mol
Boiling Pt: 192…193 °C (760 mmHg)
Melting Pt: 102…105 °C
Storage Temperature: Ambient
MDL Number: MFCD00064438
CAS Number: 624-49-7
EINECS: 210-849-0
Merck Index: 13,04308

Order Now

Specification Test Results

Appearance White to light beige to light brown Crystalline powder or crystals or flakes
Infrared spectrum Conforms
Melting point 100 to 107 °C
GC ≥98.5 %
Free acid ≤1.5 % (as Fumaric acid)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR