3-Iodoanisole 97%

Synonyms: 3-iodoanisole

AC22202-1000 AC22202-0250 AC22202-0050
103509-742EA 222 USD
103509-742 200009-990 200023-348
3-Iodoanisole 97%
3-Iodoanisole
Formula: C₇H₇IO
MW: 234.04 g/mol
Boiling Pt: 244…245 °C (760 mmHg)
Density: 1.96 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00001048
CAS Number: 766-85-8
EINECS: 212-173-1

Order Now

Specification Test Results

Appearance Clear yellow Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.6130 to 1.6150 (20 °C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR