2-Bromothiophenol 97%

Danger

Synonyms: 1-Bromo-2-mercaptobenzeneo-Bromothiophenol2-Bromobenzenethiol2-Bromobenzene-1-thiol

AC22198-0100 AC22198-0010 AC22198-0500
200004-996EA 24.9 USD
200004-996 103531-894 103531-896
2-Bromothiophenol 97%
2-Bromothiophenol
Formula: C₆H₅BrS
MW: 189.08 g/mol
Boiling Pt: 114…116 °C (10 mmHg)
Density: 1.604 g/cm³
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004827
CAS Number: 6320-02-1
EINECS: 228-665-4
UN: 2810
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.6340 to 1.6370 (20 °C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR