Diphenylamine 99+%, Extra Pure indicator grade, redox indicator

Danger

Synonyms: N-PhenylbenzeneamineN-Phenylaniline

AC22192-5000 AC22192-1000
200043-320EA 234 USD
200043-320 200043-318
Diphenylamine 99+%, Extra Pure indicator grade, redox indicator
Diphenylamine
Formula: (C₆H₅)₂NH
MW: 169.23 g/mol
Boiling Pt: 302 °C (1013 hPa)
Melting Pt: 52…54 °C
Flash Pt: 152 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00003014
CAS Number: 122-39-4
EINECS: 204-539-4
UN: 3077
ADR: 9,III
Merck Index: 13,03349
https://us.vwr.com/stibo/web/std.lang.all/76/36/2297636.jpg

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder or crystals or flakes
Infrared spectrum Conforms
Sulfated ash ≤0.05 %
GC ≥99.0 %
Sensitivity (to nitrate) Passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR