Aniline 99.5%, Extra Pure

Danger

Synonyms: PhenylamineAminobenzene

AC22173-2500 AC22173-0010 AC22173-0050 AC22173-0025
103526-918EA 200.31 USD
103526-918 200018-168 103526-920 200005-770
Aniline 99.5%, Extra Pure
Aniline
Formula: C₆H₅NH₂
MW: 93.13 g/mol
Boiling Pt: 181…185 °C (760 mmHg)
Melting Pt: –6.2 °C
Density: 1.0213 g/cm³
Flash Pt: 76 °C
Storage Temperature: Ambient
MDL Number: MFCD00007629
CAS Number: 62-53-3
EINECS: 200-539-3
UN: 1547
ADR: 6.1,II
Merck Index: 13,00661
https://us.vwr.com/stibo/web/std.lang.all/87/50/758750.jpg

Order Now

Specification Test Results

Appearance Clear colorless to yellow to orange to light brown Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.5845 to 1.5875 (20 °C, 589 nm)
Water ≤0.1 %
GC ≥99.4 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR