1,1,3,3-Tetramethylguanidine 99%
|
Danger
|
Synonyms:
N,N,N',N'-Tetramethylguanidine
AC22129-5000
200008-364EA
201
USD
200008-364
1,1,3,3-Tetramethylguanidine 99%
1,1,3,3-Tetramethylguanidine
Formula:
C₅H₁₃N₃ MW: 115.18 g/mol Boiling Pt: 160…162 °C (760 mmHg) Melting Pt: –30 °C Density: 0.91 g/cm³ Flash Pt: 50 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008323 CAS Number: 80-70-6 EINECS: 201-302-7 UN: 2920 ADR: 8,II |
Specification Test Results
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4665 to 1.4695 (20 °C, 589 nm) |
GC | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...