Gallic acid monohydrate

Warning

Synonyms: 3,4,5-Trihydroxybenzoic acid hydrate3,4,5-Trihydroxybenzoic acid monohydrate

AC22076-0025 AC22076-5000 AC22076-1000
200012-558EA 98.9 USD
200012-558 200049-564 103511-580
Gallic acid monohydrate
Gallic acid monohydrate
Formula: C₇H₆O₅·1H₂O
MW: 188.14 g/mol
Melting Pt: 251 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00149098
CAS Number: 5995-86-8
EINECS: 205-749-9

Order Now

Specification Test Results

Appearance Off-white to beige Powder
Infrared spectrum Conforms
Loss on drying ≤10 % (120 °C)
Sulfated ash ≤0.1 %
Titration with NaOH ≥99 % (On dry substance)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR