Bromoform 99+% stabilized

Danger

Synonyms: Tribromomethane

AC22069-0250 AC22069-1000 AC22069-5000
103526-876EA 211 USD
103526-876 103526-878 200057-242
Bromoform 99+% stabilized
Bromoform
Formula: Br₃CH
MW: 252.73 g/mol
Boiling Pt: 150…151 °C (760 mmHg)
Melting Pt: 8 °C
Density: 2.89 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00000128
CAS Number: 75-25-2
EINECS: 200-854-6
UN: 2515
ADR: 6.1,III
Merck Index: 13,01404
https://us.vwr.com/stibo/web/std.lang.all/78/85/717885.jpg

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
GC ≥99.0 % (excluding stabilizer)
Stabilizer ≤1.5 % w/w ethanol

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR