Methyl salicylate 99%

Warning

Synonyms: Methyl 2-hydroxybenzoateWintergreen Oil

AC22049-5000 AC22049-0025 AC22049-0010 AC22049-0050
103465-920EA 193 USD
103465-920 200013-342 200013-346 200043-934
Methyl salicylate 99%
Methyl salicylate
Formula: 2-(HO)C₆H₄CO₂CH₃
MW: 152.15 g/mol
Boiling Pt: 222 °C (760 mmHg)
Melting Pt: 8…7 °C
Density: 1.1825 g/cm³
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00002214
CAS Number: 119-36-8
EINECS: 204-317-7
Merck Index: 12,06200
https://us.vwr.com/stibo/web/std.lang.all/89/31/2298931.jpg

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5350 to 1.5380 (20 °C, 589 nm)
Density (20/4°C) 1.180 to 1.186 g/ml
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR