Acetyl chloride 99+%

Danger

AC21947-0250 AC21947-2500 AC21947-0010
200002-638EA 43.2 USD
200002-638 103526-832 200002-640
Acetyl chloride 99+%
Acetyl chloride
Formula: C₂H₃ClO
MW: 78.5 g/mol
Boiling Pt: 51 °C (760 mmHg)
Melting Pt: –112 °C
Density: 1.1 g/cm³
Flash Pt: 4 °C
Storage Temperature: Ambient
MDL Number: MFCD00000719
CAS Number: 75-36-5
EINECS: 200-865-6
UN: 1717
ADR: 3,II
Merck Index: 13,00087

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.3875 to 1.3895 (20 °C, 589 nm)
GC ≥99.0 %
Color scale ≤20 APHA
Titration RCOCl 99.0 to 101.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR