N,N-Dimethylethylamine 99%

Danger

Synonyms: DMEA

AC21929-0010 AC21929-2500
200002-632EA 105 USD
200002-632 103526-830
N,N-Dimethylethylamine 99%
N,N-Dimethylethylamine
Formula: C₄H₁₁N
MW: 73.14 g/mol
Boiling Pt: 36 °C (760 mmHg)
Melting Pt: –140 °C
Density: 0.675 g/cm³
Flash Pt: –36 °C
Storage Temperature: Ambient
MDL Number: MFCD00009039
CAS Number: 598-56-1
EINECS: 209-940-8
UN: 2733
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.3715 to 1.3735 (20 °C, 589 nm)
Titration with HClO4 ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR