2-Ethylimidazole 99%

Danger

Synonyms: 2-Ethyl-1H-imidazole

AC21928-5000 AC21928-0025 AC21928-1000
103464-602EA 266 USD
103464-602 200022-648 200034-790
2-Ethylimidazole 99%
2-Ethylimidazole
Formula: C₅H₈N₂
MW: 96.13 g/mol
Boiling Pt: 268 °C (760 mmHg)
Melting Pt: 83…87 °C
Flash Pt: 159 °C
Storage Temperature: Ambient
MDL Number: MFCD00005192
CAS Number: 1072-62-4
EINECS: 214-011-5

Order Now

Specification Test Results

Appearance White to light yellow Flakes
Infrared spectrum Conforms
Melting point 83 to 87 °C
Water ≤0.5 %
GC ≥98.5 %
GC ≤1.0 % Imidazole

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR