3-Iodopropene 97% stabilized

Danger

Synonyms: Allyl iodide3-Iodo-1-Propene3-Propenyl iodide

AC21854-1000 AC21854-0250 AC21854-5000
200018-040EA 300 USD
200018-040 200011-018 103531-852
3-Iodopropene 97% stabilized
3-Iodopropene
Formula: C₃H₅I
MW: 167.98 g/mol
Boiling Pt: 102…103 °C (760 mmHg)
Melting Pt: –99 °C
Density: 1.83 g/cm³
Flash Pt: 18 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00001094
CAS Number: 556-56-9
EINECS: 209-130-4
UN: 1723
ADR: 3,II
Merck Index: 13,00291

Order Now

Specification Test Results

Appearance Clear yellow to orange to red to brown Liquid
Infrared spectrum Conforms
Refractive index 1.5515 to 1.5565 (20 °C, 589 nm)
GC ≥96.0 %
Stabilizer Copper

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR