1-Iodonaphthalene 97.5%

Warning

AC21842-0500 AC21842-0100
200008-700EA 58.9 USD
200008-700 103509-892
1-Iodonaphthalene 97.5%
1-Iodonaphthalene
Formula: C₁₀H₇I
MW: 254.07 g/mol
Boiling Pt: 302…305 °C (760 mmHg)
Density: 1.74 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00003876
CAS Number: 90-14-2
EINECS: 201-968-9

Order Now

Specification Test Results

Appearance Clear yellow to brown Liquid
Infrared spectrum Conforms
GC ≥96.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR