1,2-Diiodobenzene 98%

Danger

AC21840-0050 AC21840-0200
200015-538EA 111 USD
200015-538 103509-890
1,2-Diiodobenzene 98%
1,2-Diiodobenzene
Formula: C₆H₄I₂
MW: 329.91 g/mol
Boiling Pt: 152 °C (15 mmHg)
Density: 2.52 g/cm³
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00001038
CAS Number: 615-42-9
EINECS: 210-425-5

Order Now

Specification Test Results

Appearance Clear yellow to brown Liquid
Infrared spectrum Conforms
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR