Aluminium nitrate nonahydrate 99+% for analysis

Danger

Synonyms: Aluminum nitrate nonahydrate

AC21828-0025
103526-802EA 185 USD
103526-802
Aluminium nitrate nonahydrate 99+% for analysis
Aluminium nitrate nonahydrate
Formula: Al(NO₃)₃·9H₂O
MW: 375.13 g/mol
Melting Pt: 73 °C
Storage Temperature: Ambient
MDL Number: MFCD00149132
CAS Number: 7784-27-2
EINECS: 236-751-8
ADR: 5.1,III

Order Now

Specification Test Results

Appearance Semitransparent to transparent Crystals
Titration Complexometric ≥99.0 %
pH 2.5 to 3.5 (5 % at 20 °C)
Chloride (Cl) ≤20 ppm
Sulfate (SO4) ≤50 ppm
Iron (Fe) ≤20 ppm
Heavy metals (as Pb) ≤20 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR