(S)-(+)-1,3-Butanediol 98%

Synonyms: (S)-(+)-1,3-Butylene glykol(S)-(+)-1,3-Dihydroxy butane(S)-Butane-1,3-diol

AC21797-0050 AC21797-0010 AC21797-0250
200032-768EA 95.1 USD
200032-768 103511-562 200032-770
(S)-(+)-1,3-Butanediol 98%
(S)-(+)-1,3-Butanediol
Formula: C₄H₁₀O₂
MW: 90.12 g/mol
Boiling Pt: 109 °C (14 mmHg)
Density: 1 g/cm³
Flash Pt: 121 °C
Storage Temperature: Ambient
MDL Number: MFCD00064278
CAS Number: 24621-61-2
EINECS: 246-363-0

Order Now

Specification Test Results

Appearance Clear colorless to yellow Viscous liquid
Infrared spectrum Conforms
Refractive index 1.4385 to 1.4405 (20 °C, 589 nm)
GC ≥97.5 %
Specific optical rotation +27° to +33° (20 °C, 589 nm) (c=1, C2H5OH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR