Hexamethylcyclotrisiloxane 98%

Danger

Synonyms: 2,2,4,4,6,6-Hexamethyl-1,3,5,2,4,6-trioxatrisilinane

AC21646-5000
103531-834EA 239 USD
103531-834
Hexamethylcyclotrisiloxane 98%
Hexamethylcyclotrisiloxane
Formula: C₆H₁₈O₃Si₃
MW: 222.46 g/mol
Boiling Pt: 134 °C (760 mmHg)
Melting Pt: 63…67 °C
Flash Pt: 35 °C
Storage Temperature: Ambient
MDL Number: MFCD00005943
CAS Number: 541-05-9
EINECS: 208-765-4
UN: 1325
ADR: 4.1,III

Order Now

Specification Test Results

Appearance Colorless to white Crystalline chunks
Infrared spectrum Conforms
Melting point 63 to 67 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR