1,4-Dichlorobutane 99%

Warning

AC21643-1000 AC21643-0010 AC21643-5000
200040-082EA 67.5 USD
200040-082 103526-766 200015-594
1,4-Dichlorobutane 99%
1,4-Dichlorobutane
Formula: ClH₂C(CH₂)₂CH₂Cl
MW: 127.01 g/mol
Boiling Pt: 161…163 °C (760 mmHg)
Melting Pt: –38 °C
Density: 1.16 g/cm³
Flash Pt: 40 °C
Storage Temperature: Ambient
MDL Number: MFCD00001011
CAS Number: 110-56-5
EINECS: 203-778-1
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4530 to 1.4550 (20 °C, 589 nm)
Water ≤0.05 %
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR