Ethanesulfonyl chloride 98+%

Danger

AC21582-0025 AC21582-0050 AC21582-0500 AC21582-5000 AC21582-1000
200019-030EA 35.4 USD
200019-030 103531-824 200043-422 200019-028 200037-318
Ethanesulfonyl chloride 98+%
Ethanesulphonyl chloride
Formula: C₂H₅ClO₂S
MW: 128.58 g/mol
Boiling Pt: 177 °C (760 mmHg)
Melting Pt: –68 °C
Density: 1.35 g/cm³
Flash Pt: 83 °C
Storage Temperature: Ambient
MDL Number: MFCD00007460
CAS Number: 594-44-5
EINECS: 209-842-5
UN: 2927
ADR: 6.1,II

Order Now

Specification Test Results

Appearance Clear colorless to yellow to light brown Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.4520 to 1.4540 (20 °C, 589 nm)
GC ≥98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR