Lignoceric acid 99%

Warning

Synonyms: Tetracosanoic acid

AC21564-2500 AC21564-0010
103511-412EA 40.2 USD
103511-412 200014-708
Lignoceric acid 99%
Lignoceric acid
Formula: C₂₄H₄₈O₂
MW: 368.64 g/mol
Boiling Pt: 272 °C (10 mmHg)
Melting Pt: 84…86 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00002810
CAS Number: 557-59-5
EINECS: 209-180-7
Merck Index: 12,05511

Order Now

Specification Test Results

Appearance White Powder or crystals or flakes
Infrared spectrum Conforms
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR