Chlormequat chloride 98%

Danger

Synonyms: (2-Chloroethyl)trimethylammonium chloride2-Chloroethyltrimethylammonium chlorideChlorocholine chloride

AC21542-1000 AC21542-0250
200033-296EA 143 USD
200033-296 200027-282
Chlormequat chloride 98%
Chlormequat chloride
Formula: ClCH₂CH₂N(CH₃)₃Cl
MW: 158.07 g/mol
Melting Pt: 239…243 °C
Storage Temperature: Ambient
MDL Number: MFCD00011869
CAS Number: 999-81-5
EINECS: 213-666-4
UN: 2811
ADR: 6.1,III
Merck Index: 13,02120

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder or crystals
Infrared spectrum Conforms
Water ≤1 % (K.F.)
Titration with HClO4 ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR