Benzalkonium chloride alkyl distribution from C8H17 to C16H33

Danger

Synonyms: Alkylbenzyldimethylammonium chlorideQuaternary ammonium compounds, benzyl-C8-18-alkyldimethyl, chlorides

AC21541-1000 AC21541-0010
200057-148EA 37.4 USD
200057-148 200057-146
Benzalkonium chloride alkyl distribution from C8H17 to C16H33
Benzalkonium chloride
Formula: C₆H₅CH₂N(CH₃)₂RCl (R=C₈H₁₇ to C₁₈H₃₇)
Density: 0.98
Storage Temperature: Ambient
MDL Number: MFCD00145757
CAS Number: 8001-54-5
EINECS: 264-151-6
UN: 1759
ADR: 8,II

Order Now

Specification Test Results

Appearance Colorless to white to yellow Solid or thick gel (May contain yellowish-white fragments)
Infrared spectrum Conforms
Assay 95.0 to 104.0 % (on anhydrous basis)
Water ≤10.0 % (K.F.)
Sulfated ash ≤0.5 %
Total amines ≤3.6 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR