Spermidine trihydrochloride 99+%

Warning

Synonyms: 4-Azoniaoctamethylenediammonium trichloride4-Azaoctamethylenediamine trihydrochlorideN1-(3-Aminopropyl)butane-1,4-diamine trihydrochlorideN-(3-Aminopropyl)-1,4-diaminobutane trihydrochloride

AC21510-0050 AC21510-0250 AC21510-0010
103511-390EA 356 USD
103511-390 200007-730 103511-388
Spermidine trihydrochloride 99+%
Spermidine trihydrochloride
Formula: C₇H₂₂Cl₃N₃
MW: 254.63 g/mol
Melting Pt: 248…252 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00012918
CAS Number: 334-50-9
EINECS: 206-379-0
Merck Index: 12,08893

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Titration Argentometric ≥99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR