4-(Trifluoromethyl)benzyl alcohol 99%

Warning

Synonyms: 4-(Hydroxymethyl)benzotrifluoride(4-(Trifluoromethyl)phenyl)methanol

AC21357-0050
200023-498EA 52.6 USD
200023-498
4-(Trifluoromethyl)benzyl alcohol 99%
4-(Trifluoromethyl)benzyl alcohol
Formula: C₈H₇F₃O
MW: 176.14 g/mol
Boiling Pt: 78…80 °C (4 mmHg)
Melting Pt: 18…20 °C
Density: 1.28 g/cm³
Flash Pt: 100 °C
MDL Number: MFCD00004661
CAS Number: 349-95-1
EINECS: 206-494-6

Order Now

Specification Test Results

Appearance Clear colorless to light brown Liquid after melting
Infrared spectrum Conforms
Refractive index 1.4590 to 1.4610 (20 °C, 589 nm)
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR