Tetrachloro-p-benzoquinone 99%
|
Warning
|
Synonyms:
Tetrachloroquinone
, p-Chloranil
, 2,3,5,6-Tetrachlorobenzo-1,4-quinone
, 2,3,5,6-Tetrachloro-1,4-benzoquinone
, Tetrachloro-1,4-benzoquinone
, 4-Chloranil
AC21356-2500
AC21356-1000
AC21356-5000
200020-834EA
416
USD
200020-834
103526-724
200026-888
Tetrachloro-p-benzoquinone 99%
Tetrachloro-p-benzoquinone
Formula:
C₆Cl₄O₂ MW: 245.88 g/mol Melting Pt: >290 °C Flash Pt: >100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001594 CAS Number: 118-75-2 EINECS: 204-274-4 UN: 2811 ADR: 6.1,III Merck Index: 13,02088 |
Specification Test Results
Appearance | Yellow to yellow-green Powder or crystals |
Infrared spectrum | Conforms |
Loss on drying | ≤0.5 % (1 g, 105 °C, 3 hrs) |
HPLC | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...