Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Manganese(IV) oxide 80-85%, Powder

Supplier: ACROS ORGANICS MS

Warning
Synonyms: Manganese dioxideManganese(IV) oxide, activatedMangan(IV)-oxidManganese peroxide

AC21349-0025 AC21349-0250 AC21349-0010
103509-616EA 149.03 USD InStock
103509-616 200013-126 200019-592
Manganese(IV) oxide 80-85%, Powder
Manganese (IV) oxide


Formula: MnO₂
MW: 86.94 g/mol
Melting Pt: 535 °C
Density: 5.03 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003463
CAS Number: 1313-13-9
EINECS: 215-202-6
Merck Index: 13,05753
Order Now
Specification Test Results
Appearance Dark grey Powder
Loss on drying ≤0.5 % (105°C)
Titration with Na2S2O3 ≥80 %
Silicate (as SiO2) ≤7 %
Iron (Fe) ≤1 %
Insoluble matter ≤10 % (in HCl)


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty