Nitromethane 99+% for analysis

Warning

AC21337-0025 AC21337-5000 AC21337-1000
200002-516EA 115.38 USD
200002-516 200013-684 103526-716
Nitromethane 99+% for analysis
Nitromethane
Formula: CH₃NO₂
MW: 61.04 g/mol
Boiling Pt: 100…102 °C (760 mmHg)
Melting Pt: –29 °C
Density: 1.12 g/cm³
Flash Pt: 35 °C
Storage Temperature: Ambient
MDL Number: MFCD00007400
CAS Number: 75-52-5
EINECS: 200-876-6
UN: 1261
ADR: 3,II
Merck Index: 12,06708

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.3805 to 1.3825 (20 °C, 589 nm)
Water ≤0.5 % (Coulometric)
GC ≥99.0 %
Residue after evaporation ≤0.005 %
Cadmium (Cd) ≤0.1 ppm
Copper (Cu) ≤0.1 ppm
Iron (Fe) ≤0.5 ppm
Lead (Pb) ≤0.1 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR