Mercury(I) chloride 99+%, Extra Pure

Danger

Synonyms: Mercurous chlorideDimercury dichlorideCalomel

AC21312-5000
103531-808EA 334 USD
103531-808
Mercury(I) chloride 99+%, Extra Pure
Mercury (I) chloride
Formula: Cl₂Hg₂
MW: 472.09 g/mol
Boiling Pt: 384 °C (1013 hPa)
Melting Pt: 400 °C (subl.)
Storage Temperature: Ambient
MDL Number: MFCD00011043
CAS Number: 10112-91-1
EINECS: 233-307-5
UN: 2025
ADR: 6.1,III
Merck Index: 12,05951

Order Now

Specification Test Results

Appearance Off-white Powder
Titration Iodimetric ≥99.0 %
Mercuric Chloride ≤0.01 % (HgCl2)
Residue after reduction ≤0.02 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR