New fuchsine

Warning

Synonyms: Basic Violet 2New Fuchsin4-[(4-Amino-m-tolyl)(4-imino-3-methylcyclohexa-2,5-dien-1-ylidene)methyl]-o-toluidine hydrochloride

AC21214-0250 AC21214-1000
103509-816EA 139 USD
103509-816 200032-214
New fuchsine
New fuchsine
Formula: C₂₂H₂₄ClN₃
MW: 365.91 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00012567
CAS Number: 3248-91-7
EINECS: 221-831-7

Order Now

Specification Test Results

Appearance Dark green Crystalline powder or crystals
Infrared spectrum Conforms
UV Lambda max 550 to 554 nm (in 50% ethanol)
Absorption Ratio P-15/P+15 1.1 to 1.4
Specific absorbance A (1%/1cm) ≥1500 (at lambda max.)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR