Benzyltrimethylammonium chloride 98+%

Danger

Synonyms: N,N,N-Trimethyl-1-phenylmethanaminium chloride

AC21186-2500 AC21186-0010 AC21186-0500
200005-832EA 52.1 USD
200005-832 103524-148 102773-858
Benzyltrimethylammonium chloride 98+%
Benzyltrimethylammonium chloride
Formula: C₁₀H₁₆ClN
MW: 185.7 g/mol
Melting Pt: 239 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00011782
CAS Number: 56-93-9
EINECS: 200-300-3
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to cream Crystalline powder and/or chunks
Infrared spectrum Conforms
Titration Argentometric ≥98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR