Magnesium acetate tetrahydrate ≥97.5%, crystalline, Extra Pure

Supplier: ACROS ORGANICS MS

Synonyms: Magnesium di(acetate) tetrahydrateAcetic acid magnesium salt tetrahydrate

AC21157-2500 AC21157-0010 AC21157-0051
200013-086EA 58.91 USD InStock
200013-086 200019-548 103511-332
Magnesium acetate tetrahydrate ≥97.5%, crystalline, Extra Pure
Magnesium acetate tetrahydrate
Formula: (H₃CCOO)₂Mg·4H₂O
MW: 214.46 g/mol
Melting Pt: 80 °C
Density: 1.454 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149214
CAS Number: 16674-78-5
EINECS: 205-554-9
Merck Index: 13,05676

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Titration Complexometric ≥97 %
Chloride (Cl) ≤100 ppm
Sulfate (SO4) ≤500 ppm
Iron (Fe) ≤50 ppm
Heavy metals (as Pb) ≤20 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR