3-Chloro-4-fluoroaniline 98%

Warning

AC21153-1000 AC21153-0250
200035-312EA 124 USD
200035-312 200016-990
3-Chloro-4-fluoroaniline 98%
3-Chloro-4-fluoroaniline
Formula: C₆H₅ClFN
MW: 145.56 g/mol
Boiling Pt: 230 °C (1013 hPa)
Melting Pt: 42…47 °C
Flash Pt: 149 °C
Storage Temperature: Ambient
MDL Number: MFCD00007767
CAS Number: 367-21-5
EINECS: 206-682-8
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Beige to brown Crystalline solid (May darken on storage)
Infrared spectrum Conforms
Melting point 42 to 47 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR