Calcium-D gluconate 99%

Synonyms: Calcium (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoateCalcium GluconateD-Gluconic acid calcium salt

AC21106-0010 AC21106-2500 AC21106-0051
200018-710EA 42.7 USD
200018-710 200018-708 103516-760
Calcium-D gluconate 99%
Calcium D-gluconate
Formula: HOH₂C(CH(OH))₄COOCa
MW: 430.38 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00064209
CAS Number: 299-28-5
EINECS: 206-075-8
Merck Index: 13,01671

Order Now

Specification Test Results

Appearance White to off-white Crystalline or granular powder
Infrared spectrum Conforms
Loss on drying ≤2.0 % (1 g, 105 °C, 16 hrs)
Heavy metals ≤20 ppm
Titration after Ion exchange 98.5 to 102.0 % (as C12H22CaO14.H2O)
Chloride (Cl) ≤700 ppm
Sulfate (SO4) ≤500 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR