Benzyl isocyanate 99+%

Danger

AC21076-0250 AC21076-0050
200018-264EA 249 USD
200018-264 200005-824
Benzyl isocyanate 99+%
Benzyl isocyanate
Formula: C₈H₇NO
MW: 133.15 g/mol
Boiling Pt: 101…104 °C (33 mmHg)
Density: 1.07 g/cm³
Flash Pt: 45 °C
MDL Number: MFCD00009701
CAS Number: 3173-56-6
EINECS: 221-640-9
UN: 3080
ADR: 6.1,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5241 to 1.5261 (20 °C, 589 nm)
GC ≥99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR