3-Chloroacetylacetone 98%

Warning

Synonyms: 3-Chloropentane-2,4-dione3-Chloro-2,4-pentanedione

AC21067-5000 AC21067-1000 AC21067-0050
200023-224EA 52.8 USD
200023-224 200029-232 102776-736
3-Chloroacetylacetone 98%
3-Chloroacetylacetone
Formula: C₅H₇ClO₂
MW: 134.56 g/mol
Boiling Pt: 49…52 °C (18 mmHg)
Melting Pt: –15 °C
Density: 1.179 g/cm³
Flash Pt: 56 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00009651
CAS Number: 1694-29-7
EINECS: 216-902-4
UN: 1224
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear yellow to brown Liquid
Infrared spectrum Conforms
Refractive index 1.4825 to 1.4845 (20 °C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR